JWPH37C050AC J & W Pharmlab LLC

Methyl 5-bromo-4-trifluoromethyl-thiazole-2-carboxylate

  • MDL number MFCD11506343
  • Molecular Weight : 290.0544

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link https://www.jwpharmlab.com/home/product/90R0246
Availability details for
Quantity
  • Availability
  • Add to Cart