JWPH37C050AF J & W Pharmlab LLC

3-(2-Amino-ethyl)-4-bromo-phenol hydrochloride

  • MDL number MFCD31736185
  • Molecular Weight : 252.538

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link https://www.jwpharmlab.com/home/product/20R0989S
Availability details for
Quantity
  • Availability
  • Add to Cart