JWPH37C050B0 J & W Pharmlab LLC

Benzo[1,3]dioxole-5-carbaldehyde oxime

  • MDL number MFCD00016905
  • Cas Number : 2089-36-3
  • Molecular Weight : 165.1482

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form Light Yellow Solid
Shipping Temperature RT
Country of Origin USA
Product Link https://www.jwpharmlab.com/home/product/10R0732
Availability details for
Quantity
  • Availability
  • Add to Cart