JWPH37C050B6 J & W Pharmlab LLC

4,4-Difluoro-cyclohexanone oxime

  • MDL number MFCD31543944
  • Molecular Weight : 149.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link https://www.jwpharmlab.com/home/product/10R0731
Availability details for
Quantity
  • Availability
  • Add to Cart