JWPH37C050B7 J & W Pharmlab LLC

(2,4-Dimethoxy-phenyl)-carbamic acid tert-butyl ester

  • MDL number MFCD21170312
  • Molecular Weight : 253.2974

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link https://www.jwpharmlab.com/home/product/03R0397
Availability details for
Quantity
  • Availability
  • Add to Cart