JWPH37C050BD J & W Pharmlab LLC

[(1-tert-Butoxycarbonylamino-cyclopropanecarbonyl)-amino]-acetic acid ethyl ester

  • MDL number MFCD32067016
  • Cas Number : 129306-15-6
  • Molecular Weight : 286.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Physical Form Off White Solid
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/10R0733

Safety Information

MSDS https://www.jwpharmlab.com/home/product/10R0733
Availability details for
Quantity
  • Availability
  • Add to Cart