JWPH57C9E24B J & W Pharmlab LLC

4-Chloromethyl-oxazole-2-carboxylic acid ethyl ester

  • MDL number MFCD32066988
  • Cas Number : 2376143-24-5
  • Molecular Weight : 189.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Physical Form Light Yellow Liquid
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/56R0651

Safety Information

MSDS https://www.jwpharmlab.com/home/product/56R0651
Availability details for
Quantity
  • Availability
  • Add to Cart