JWPH57C9E26F J & W Pharmlab LLC

3-Fluoro-cyclohexylamine

  • MDL number MFCD18791204
  • Cas Number : 1273566-51-0
  • Molecular Weight : 117.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/20R1704

Safety Information

MSDS https://www.jwpharmlab.com/home/product/20R1704
Availability details for
Quantity
  • Availability
  • Add to Cart