JWPH57C9E287 J & W Pharmlab LLC

(2ALPHA,3ABETA,5ALPHA,6ABETA)-OCTAHYDRO-2,5-PENTALENEDIAMINE

  • MDL number MFCD28390115
  • Cas Number : 197015-83-1
  • Molecular Weight : 140.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Physical Form Light Yellow Solid
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/20R1757

Safety Information

MSDS https://www.jwpharmlab.com/home/product/20R1757
Availability details for
Quantity
  • Availability
  • Add to Cart