JWPH57C9E292 J & W Pharmlab LLC

(S)-4-Boc-6-Amino-[1,4]oxazepane

  • MDL number MFCD32182545
  • Cas Number : 1932377-93-9
  • Molecular Weight : 216.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form Light Yellow Liquid
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/53R0219

Safety Information

MSDS https://www.jwpharmlab.com/home/product/53R0219
Availability details for
Quantity
  • Availability
  • Add to Cart