JWPH580E1311 J & W Pharmlab LLC

(R)-5-Bromo-3-methyl-2,3-dihydro-isoindol-1-one

  • Molecular Weight : 226.0723

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.96
Country of Origin USA
Product Link https://www.jwpharmlab.com/home/product/48R0901
Availability details for
Quantity
  • Availability
  • Add to Cart