JWPH580E1312 J & W Pharmlab LLC

N-,N-diboc-Oxazol-2-yl-methylamine

  • Molecular Weight : 298.3382

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.96
Country of Origin USA
Product Link https://www.jwpharmlab.com/home/product/56R0655
Availability details for
Quantity
  • Availability
  • Add to Cart