JWPH580E1314 J & W Pharmlab LLC

3-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-pyrrolo[1,2-a]pyrimidine-6-carboxylic acid ethyl ester

  • Molecular Weight : 316.1629

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.96
Country of Origin USA
Product Link https://www.jwpharmlab.com/home/product/70R0535P
Availability details for
Quantity
  • Availability
  • Add to Cart