JWPH580E131A J & W Pharmlab LLC

5-Benzyl-2-oxa-5-aza-bicyclo[2.2.2]octane-4-carboxylic acid

  • MDL number na
  • Molecular Weight : 247.2932

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin USA
Product Link https://www.jwpharmlab.com/home/product/53R0281
Availability details for
Quantity
  • Availability
  • Add to Cart