JWPH580E132F J & W Pharmlab LLC

(R)-2,3-Dihydro-benzofuran-3-ylamine

  • MDL number MFCD06761955
  • Molecular Weight : 135.1652

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.96
Country of Origin USA
Product Link https://www.jwpharmlab.com/home/product/20R0158
Availability details for
Quantity
  • Availability
  • Add to Cart