JWPH580E1341 J & W Pharmlab LLC

trans-[2-(4-Fluoro-phenyl)-cyclopropyl]-carbamic acid tert-butyl ester

  • MDL number na
  • Molecular Weight : 251.3001

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Physical Form White Solid
Country of Origin USA
Product Link https://www.jwpharmlab.com/home/product/20R1776
Availability details for
Quantity
  • Availability
  • Add to Cart