JWPH580E1343 J & W Pharmlab LLC

(3S)-2,3-Dihydro-benzofuran-3-ylamine

  • MDL number MFCD06761956
  • Cas Number : 1228569-93-4
  • Molecular Weight : 135.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/20R1779

Safety Information

MSDS https://www.jwpharmlab.com/home/product/20R1779
Availability details for
Quantity
  • Availability
  • Add to Cart