JWPH580E1351 J & W Pharmlab LLC

4-Chloromethyl-oxazole-2-carboxylic acid methyl ester

  • MDL number MFCD22544011
  • Cas Number : 1393584-98-9
  • Molecular Weight : 175.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Physical Form Off White Solid
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/56R0571

Safety Information

MSDS https://www.jwpharmlab.com/home/product/56R0571
Availability details for
Quantity
  • Availability
  • Add to Cart