JWPH580E135A J & W Pharmlab LLC

TERT-BUTYL 4-BROMOAZEPANE-1-CARBOXYLATE

  • MDL number MFCD31667058
  • Molecular Weight : 278.1885

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.96
Country of Origin USA
Product Link https://www.jwpharmlab.com/home/product/60R1424
Availability details for
Quantity
  • Availability
  • Add to Cart