JWPH580E1362 J & W Pharmlab LLC

(3-Bromo-phenyl)-ethyl-methyl-amine

  • MDL number MFCD23708966
  • Molecular Weight : 214.1045

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.96
Country of Origin USA
Product Link https://www.jwpharmlab.com/home/product/69R1384
Availability details for
Quantity
  • Availability
  • Add to Cart