JWPH580E136E J & W Pharmlab LLC

3-Methyl-pyrrolidine-1,3-dicarboxylic acid 1-benzyl ester

  • MDL number MFCD18274759
  • Molecular Weight : 263.2926

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Physical Form White Solid
Country of Origin USA
Product Link https://www.jwpharmlab.com/home/product/75R0548
Availability details for
Quantity
  • Availability
  • Add to Cart