JWPH580E1370 J & W Pharmlab LLC

5-Trifluoromethyl-pyrrolidine-2-carboxylic acid

  • MDL number MFCD28389194
  • Molecular Weight : 283.2469

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Physical Form White Solid
Country of Origin USA
Product Link https://www.jwpharmlab.com/home/product/75R0887
Availability details for
Quantity
  • Availability
  • Add to Cart