JWPH580E137B J & W Pharmlab LLC

(R)-2,3-Dihydro-benzofuran-3-ylamine hydrochloride

  • MDL number MFCD28986152
  • Molecular Weight : 171.6261

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.96
Country of Origin USA
Product Link https://www.jwpharmlab.com/home/product/20R1778S
Availability details for
Quantity
  • Availability
  • Add to Cart