JWPH580E1386 J & W Pharmlab LLC

4-(2-Bromo-1,1-dimethyl-ethyl)-2,2-dimethyl-[1,3]dioxolane

  • MDL number na
  • Molecular Weight : 237.1361

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Physical Form 0
Country of Origin USA
Product Link https://www.jwpharmlab.com/home/product/05R0955
Availability details for
Quantity
  • Availability
  • Add to Cart