JWPH580E139A J & W Pharmlab LLC

1H-Pyrazolo[3,4-c]pyridine-3-carboxylic acid amide

  • MDL number MFCD23703881
  • Molecular Weight : 162.1506

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin USA
Product Link https://www.jwpharmlab.com/home/product/45R0904
Availability details for
Quantity
  • Availability
  • Add to Cart