JWPH74A3E180 J & W Pharmlab LLC

tert-Butoxycarbonylamino-cyclohexanecarboxylic acid ethyl ester

  • MDL number MFCD24444907
  • Cas Number : 1400583-09-6
  • Molecular Weight : 271.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/15R0583

Safety Information

MSDS https://www.jwpharmlab.com/home/product/15R0583
Availability details for
Quantity
  • Availability
  • Add to Cart