JWPH868A68DA J & W Pharmlab LLC

N-(2-Formyl-phenyl)-4-methyl-benzenesulfonamide

  • MDL number MFCD00805186
  • Cas Number : 6590-65-4
  • Molecular Weight : 275.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form Off White Solid
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/10R0747

Safety Information

MSDS https://www.jwpharmlab.com/home/product/10R0747
Availability details for
Quantity
  • Availability
  • Add to Cart