JWPH93D8B456 J & W Pharmlab LLC

endo-1-Aza-bicyclo[2.2.1]hept-3-ylamine dihydrochloride

  • MDL number MFCD32664163
  • Molecular Weight : 185.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3S,4S)-1-azabicyclo[2.2.1]heptan-3-amine dihydrochloride

Description

Purity 98%
Physical Form White Solid
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/75R0949S

Safety Information

MSDS https://www.jwpharmlab.com/home/product/75R0949S
Availability details for
Quantity
  • Availability
  • Add to Cart