JWPH93D8B46A J & W Pharmlab LLC

2-Trifluoromethyl-2H-pyrazole-3-carbaldehyde

  • MDL number MFCD30188195
  • Cas Number : 2091369-84-3
  • Molecular Weight : 164.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/68R0659

Safety Information

MSDS https://www.jwpharmlab.com/home/product/68R0659
Availability details for
Quantity
  • Availability
  • Add to Cart