JWPH93D8B4B3 J & W Pharmlab LLC

1-(4-Bromo-isothiazol-5-yl)-ethanone

  • MDL number MFCD32068943
  • Cas Number : 1780693-60-8
  • Molecular Weight : 206.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(4-bromoisothiazol-5-yl)ethan-1-one
Inchi Code 1S/C5H4BrNOS/c1-3(8)5-4(6)2-7-9-5/h2H,1H3
Inchi Key BEEDUSXMYDFQDF-UHFFFAOYSA-N

Description

Purity 97%
Physical Form Off White Solid
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/90R0488

Safety Information

MSDS https://www.jwpharmlab.com/home/product/90R0488
Availability details for
Quantity
  • Availability
  • Add to Cart