JWPH93E485D8 J & W Pharmlab LLC

(1S,3S)-(3-Hydroxy-cycloheptyl)-carbamic acid tert-butyl ester

  • Cas Number : 1403864-29-8
  • Molecular Weight : 229.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/03R0466

Safety Information

MSDS https://www.jwpharmlab.com/home/product/03R0466
Availability details for
Quantity
  • Availability
  • Add to Cart