JWPH93E48607 J & W Pharmlab LLC

6-Oxa-bicyclo[3.2.1]oct-3-en-7-one

  • MDL number MFCD21091252
  • Cas Number : 4720-83-6
  • Molecular Weight : 124.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Physical Form Pale Yellow Oil
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/05R0973

Safety Information

MSDS https://www.jwpharmlab.com/home/product/05R0973
Availability details for
Quantity
  • Availability
  • Add to Cart