JWPH942F3141 J & W Pharmlab LLC

3-(2-Chloro-phenyl)-3-oxo-propionic acid tert-butyl ester

  • MDL number MFCD09832164
  • Cas Number : 263239-24-3
  • Molecular Weight : 254.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/05R0296

Safety Information

MSDS https://www.jwpharmlab.com/home/product/05R0296
Availability details for
Quantity
  • Availability
  • Add to Cart