JWPH942F319D J & W Pharmlab LLC

(4,5-Dimethoxy-2-nitro-phenyl)-acetic acid methyl ester

  • MDL number MFCD00015617
  • Cas Number : 2982-53-8
  • Molecular Weight : 255.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/05R0354

Safety Information

MSDS https://www.jwpharmlab.com/home/product/05R0354
Availability details for
Quantity
  • Availability
  • Add to Cart