JWPH942F31AB J & W Pharmlab LLC

(2-Chloro-4-nitro-phenyl)-acetonitrile

  • MDL number MFCD00024318
  • Cas Number : 89277-99-6
  • Molecular Weight : 196.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/05R0448

Safety Information

MSDS https://www.jwpharmlab.com/home/product/05R0448
Availability details for
Quantity
  • Availability
  • Add to Cart