JWPH942F31C4 J & W Pharmlab LLC

(3-Chloro-phenyl)-cyano-acetic acid ethyl ester

  • MDL number MFCD03412570
  • Cas Number : 92847-34-2
  • Molecular Weight : 223.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/05R0487

Safety Information

MSDS https://www.jwpharmlab.com/home/product/05R0487
Availability details for
Quantity
  • Availability
  • Add to Cart