JWPH942F31C7 J & W Pharmlab LLC

2-Oxo-2,3-dihydro-benzofuran-4-carboxylic acid

  • MDL number MFCD18914306
  • Cas Number : 199122-01-5
  • Molecular Weight : 178.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/05R0498

Safety Information

MSDS https://www.jwpharmlab.com/home/product/05R0498
Availability details for
Quantity
  • Availability
  • Add to Cart