JWPH942F31C8 J & W Pharmlab LLC

1-Oxo-1,3-dihydro-isobenzofuran-4-carboxylic acid

  • MDL number MFCD15144243
  • Cas Number : 4792-27-2
  • Molecular Weight : 178.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/05R0499

Safety Information

MSDS https://www.jwpharmlab.com/home/product/05R0499
Availability details for
Quantity
  • Availability
  • Add to Cart