JWPH942F31F1 J & W Pharmlab LLC

cis-Cyclobutane-1,3-dicarboxylic acid monobenzyl ester

  • MDL number MFCD09029032
  • Cas Number : 1638764-75-6
  • Molecular Weight : 234.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/05R0687

Safety Information

MSDS https://www.jwpharmlab.com/home/product/05R0687
Availability details for
Quantity
  • Availability
  • Add to Cart