JWPH942F3203 J & W Pharmlab LLC

3-Bromo-4-chloro-2-fluoro-benzoic acid

  • MDL number MFCD28398041
  • Cas Number : 1677706-23-8
  • Molecular Weight : 253.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/05R0746

Safety Information

MSDS https://www.jwpharmlab.com/home/product/05R0746
Availability details for
Quantity
  • Availability
  • Add to Cart