JWPH942F323D J & W Pharmlab LLC

4,5-Dimethyl-phthalonitrile

  • MDL number MFCD28964903
  • Cas Number : 36360-43-7
  • Molecular Weight : 156.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/05R0900

Safety Information

MSDS https://www.jwpharmlab.com/home/product/05R0900
Availability details for
Quantity
  • Availability
  • Add to Cart