JWPH942F3241 J & W Pharmlab LLC

3-Methoxycarbonyl-cyclopentane-boronic acid

  • MDL number MFCD32066999
  • Cas Number : 2408428-26-0
  • Molecular Weight : 171.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/05R0921

Safety Information

MSDS https://www.jwpharmlab.com/home/product/05R0921
Availability details for
Quantity
  • Availability
  • Add to Cart