JWPH942F3262 J & W Pharmlab LLC

4'-Chloro-4-fluoro-3-nitrobenzophenone

  • MDL number MFCD04117956
  • Cas Number : 956474-49-0
  • Molecular Weight : 279.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/10R0509

Safety Information

MSDS https://www.jwpharmlab.com/home/product/10R0509
Availability details for
Quantity
  • Availability
  • Add to Cart