JWPH942F326D J & W Pharmlab LLC

4-Formyl-indan-1-one 1,2-ethanediol ketal

  • MDL number MFCD09701206
  • Cas Number : 954238-31-4
  • Molecular Weight : 204.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/10R0178

Safety Information

MSDS https://www.jwpharmlab.com/home/product/10R0178
Availability details for
Quantity
  • Availability
  • Add to Cart