JWPH942F328A J & W Pharmlab LLC

3-Methoxy-5-trifluoromethyl-benzoic acid

  • MDL number MFCD09025409
  • Cas Number : 53985-48-1
  • Molecular Weight : 220.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/10R0372

Safety Information

MSDS https://www.jwpharmlab.com/home/product/10R0372
Availability details for
Quantity
  • Availability
  • Add to Cart