JWPH942F3290 J & W Pharmlab LLC

6-Trifluoromethyl-1H-benzoimidazole-2-carbaldehyde

  • MDL number MFCD18432486
  • Cas Number : 944903-91-7
  • Molecular Weight : 214.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/10R0395

Safety Information

MSDS https://www.jwpharmlab.com/home/product/10R0395
Availability details for
Quantity
  • Availability
  • Add to Cart