JWPH942F32C6 J & W Pharmlab LLC

3,4,5-Trifluoro-benzamidine

  • MDL number MFCD05663401
  • Cas Number : 504404-35-7
  • Molecular Weight : 174.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/12R0070

Safety Information

MSDS https://www.jwpharmlab.com/home/product/12R0070
Availability details for
Quantity
  • Availability
  • Add to Cart