JWPH942F32D6 J & W Pharmlab LLC

1-(4-Amino-2-fluoro-phenyl)-ethanone

  • MDL number MFCD08234548
  • Cas Number : 112279-56-8
  • Molecular Weight : 153.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/10R0651

Safety Information

MSDS https://www.jwpharmlab.com/home/product/10R0651
Availability details for
Quantity
  • Availability
  • Add to Cart