JWPH942F3302 J & W Pharmlab LLC

2,4-Dimethyl-benzamidine hydrochloride

  • MDL number MFCD04114462
  • Cas Number : 1187927-39-4
  • Molecular Weight : 184.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/12R0080S

Safety Information

MSDS https://www.jwpharmlab.com/home/product/12R0080S
Availability details for
Quantity
  • Availability
  • Add to Cart