JWPH942F330C J & W Pharmlab LLC

1H-Indole-5-carboxamidine hydrochloride

  • MDL number MFCD09832173
  • Cas Number : 154022-28-3
  • Molecular Weight : 195.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. RT
Shipping Temperature RT
Country of Origin US
Product Link https://www.jwpharmlab.com/home/product/12R0021S

Safety Information

MSDS https://www.jwpharmlab.com/home/product/12R0021S
Availability details for
Quantity
  • Availability
  • Add to Cart